Learn More
Apexbio Technology LLC ACSF,25ml
Supplier: Apexbio Technology LLC B544325
ACSF is often used as replacement of CSF for perfusion of brain slices to preserve interneurons [1].ACSF (artificial cerebrospinal fluid) is invented to reduce the incidence of cerebral edema and further suppress brain cell disorders. And ACSF is often used as an irrigation fluid or perfusion fluid in the field of neurosurgery, such as intracranial surgery, or when used as a replenishing fluid for lost cerebrospinal fluid. ACSF is also used to maintain pH balance and tissue oxygen delivery for research [2, 3]. Other sizes are also available. Please inqury us for quote.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.