Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACT 335827, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and selective orexin OX1 receptor antagonist (Kb values are 41 and 560 nM for OX1 and OX2 receptors respectively). Elicits anxiolytic effects in vivo. Brain penetrant and orally available.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 518.64 |
Chemical Name or Material | ACT 335827 |
Safety and Handling
Recommended Storage | Store at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction