Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ ACVR1 (R206H) (Human) Recombinant Protein
Shop All Abnova Corporation Products
Click to view available options
Quantity:
5 μg
Description
Human ACVR1 (NP_001096.1, 145 a.a. - 509 a.a.) R206H mutant partial recombinant protein with GST-tag at N-terminal using baculovirus expression system.
Specifications
Specifications
For Use With (Application) | Functional Study |
Gene ID (Entrez) | 90 |
Name | activin A receptor, type I |
Quality Control Testing | The purity was assessed by SDS-PAGE/CBB staining. |
Quantity | 5 μg |
Storage Requirements | Stored at -80°C. Aliquot to avoid repeated freezing and thawing. |
Gene Alias | ACTRI, ACVR1A, ACVRLK2, ALK2, FOP, SKR1, TSRI |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction