Learn More
Seca Corp seca 437 Adapter for flat scales seca 869, 874, and 876 in conjunction with seca 217 stadiometer
Adapter for flat scales seca 869, 874 / 874dr, and 876 in conjunction with seca 217 stadiometer.With the help of the adapter element seca 437, the stadiometer seca 217 can be attached to the flat scales seca 869, seca 874 or seca 876 to make a perfect weighing and measuring station for school children and adults. The result is two measurements of best quality in just one step. Even though the elements are lightweight and easily assembled, the system is extremely stable and warp-resistant. Almost like a permanent installation.• Converts the seca 217 and a flat scale in a complete mobile measuring station• An integrated click system secures a stable and safety weighing process• Easy and fast assembly• High-quality: made of robust plasticWeight: 1.3 lbs / 0.60 kgWidth: 13.1 inch / 33.3 cmHeight: 3.4 inch / 8.6 cmDepth: 12.2 inch / 31 cm