Learn More
TARGETMOL CHEMICALS INC ADEFOVIR DIPIVOXIL 500MG
Supplier:  TARGETMOL CHEMICALS INC T1675
Encompass_Preferred
Also available in 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Adefovir dipivoxil (GS 0840) is a dipivoxil formulation of adefovir a nucleoside reverse transcriptase inhibitor analog of adenosine with activity against hepatitis B virus (HBV) herpes virus and human immunodeficiency virus (HIV). purity: 98%
Catalog No. 50-258-1158
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.