Learn More
AARON CHEMICALS LLC ADENOSINE-3,5-CYCLIC-25MG
Supplier:  AARON CHEMICALS LLC AR003CJJ25MG
Encompass
Adenosine-3',5'-cyclic monophosphate monosodium salt, 25MG, CAS# 37839-81-9, MFCD00150890, MOLWT: 352.19
Catalog No. NC2308357
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.