Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Adenosine-3',5'-cyclic monophosphate sodium salt, 99%

Catalog No. p-7042335
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 37839-81-9 | C10H11N5NaO6P | 351.19 g/mol

Adenosine 3?,5?-cyclic monophosphate sodium salt monohydrate is used as an activator of protein kinase A (PKA) . A key regulator of many cellular reactions. Released in response to hormonal stimulation. . Directly activates Epac, a Rap1 guanine-nucleotide exchange factor. Acts as a second messenger in signal transduction mechanisms. Promotes apoptosis in resting human B lymphocytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adenosine 3′,5′-cyclic monophosphate sodium salt monohydrate is used as an activator of protein kinase A (PKA) . A key regulator of many cellular reactions. Released in response to hormonal stimulation. . Directly activates Epac, a Rap1 guanine-nucleotide exchange factor. Acts as a second messenger in signal transduction mechanisms. Promotes apoptosis in resting human B lymphocytes.

Solubility
Soluble in water (50 mg/ml).

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 37839-81-9
Molecular Formula C10H11N5NaO6P
Molecular Weight (g/mol) 351.19
MDL Number MFCD00069736
InChI Key BXJBFCKTIWRKMQ-GWKNMROSNA-M
Synonym adenosine 3',5'-cyclic monophosphate sodium salt monohydrate, c10h11n5o6p.na.h2o, sodium 3',5'-cyclic amp anion hydrate, adenosine 3',5'-cyclic monophosphate monosodium salt
PubChem CID 91872444
IUPAC Name sodium;(4aR,6R,7R,7aS)-6-(6-aminopurin-9-yl)-2-oxido-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-7-ol;hydrate
SMILES [Na+].NC1=C2N=CN([C@@H]3O[C@@H]4COP([O-])(=O)O[C@H]4[C@H]3O)C2=NC=N1

Specifications

Color White
Quantity 250 mg
Beilstein 54612
Solubility Information Soluble in water (50mg/ml).
Formula Weight 351.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Adenosine-3',5'-cyclic monophosphate sodium salt
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.