Learn More
American Radiolabeled Chemicals Inc ADENOSINE 5 8-3H 1LC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 18121MC
Encompass_Preferred
Adenosine-5',8-3H 1mCi in ethanol:water (1:1), 1 mCi/ml, S.A. 60-90 Ci/mmol, M.W. 267.24, Shipped in Dry Ice
Catalog No. 50-901-231
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.