Learn More
Medchemexpress LLC HY-W010918 100mg Medchemexpress, Adenosine 5'-diphosphate CAS:58-64-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW010918100MG
Encompass_Preferred
Medchemexpress, HY-W010918 100mg Adenosine 5'-diphosphate CAS:58-64-0 Adenosine 5'-diphosphate is a nucleoside diphosphate. Adenosine 5'-diphosphate is the product of ATP dephosphorylation by ATPases. Adenosine 5'-diphosphate induces human platelet aggregation and inhibits stimulated adenylate cyclase by an action at P 2T -purinoceptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4544
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.