Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine-5'-Diphosphate Trisodium Salt, MP Biomedicals

Catalog No. p-3179269 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 g
5 g
10 g

Chemical Identifiers

CAS 2092-65-1
Molecular Formula C10H15N5Na3O10P2
Molecular Weight (g/mol) 496.172
InChI Key VJZLETZZUNQROW-MSQVLRTGSA-N
Synonym adenosine-5'-diphosphate trisodium salt
PubChem CID 131875859
IUPAC Name [(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphono hydrogen phosphate;sodium
SMILES C1=NC2=C(C(=N1)N)N=CN2C3C(C(C(O3)COP(=O)(O)OP(=O)(O)O)O)O.[Na].[Na].[Na]

Specifications

Quantity 1 g
Formula Weight 493.2
Chemical Name or Material Adenosine-5'-Diphosphate Trisodium Salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.