Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine 5'-monophosphate monohydrate, 99.7%, MP Biomedicals™

Catalog No. p-4403616 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 g
25 g

Used as a monomer in RNA. It is an ester of phosphoric acid and the nucleoside adenosine. AMP consists of a phosphate group, the sugar ribose, and the nucleobase adenine

Chemical Identifiers

CAS 18422-05-4
Molecular Formula C10H16N5O8P
Molecular Weight (g/mol) 365.24
MDL Number MFCD00005750,MFCD00149360,MFCD00005750,MFCD00149360
InChI Key ZOEFQKVADUBYKV-MCDZGGTQSA-N
Synonym 5'-Adenylic acid, 5'-AMP
PubChem CID 122130273
IUPAC Name {[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}phosphonic acid hydrate
SMILES O.NC1=C2N=CN([C@@H]3O[C@H](COP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Color White
Assay Percent Range 99.70%
Quantity 1 g
Formula Weight 347.2
Physical Form Crystalline Powder
Chemical Name or Material Adenosine 5'-monophosphate monohydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.