Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine-5'-triphosphate disodium salt tryhydrate, ≥96%, MP Biomedicals™

Catalog No. p-4404649 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
10 g
25 g

Chemical Identifiers

CAS 51963-61-2
Molecular Formula C10H20N5Na2O16P3
Molecular Weight (g/mol) 605.19
MDL Number MFCD00150754
InChI Key MWEQTWJABOLLOS-MVHBFMHHNA-L
Synonym adenosine 5'-triphosphate disodium salt trihydrate, adenosine 5'-triphosphate disodium salt, adenosine 5'-triphosphate disodium salt trihydrate, disodium salt
PubChem CID 134129625
IUPAC Name disodium trihydrate hydrogen [({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]phosphonate
SMILES O.O.O.[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Color White
Physical Form Crystalline Powder
Quantity 10 g
Assay Percent Range ≥96%
Formula Weight 551.2 (Anhydrous)
Chemical Name or Material Adenosine-5'-triphosphate disodium salt tryhydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.