Learn More
Revvity Health Sciences Inc Adenosine A1R fluorescent probe, 1 Unit
Supplier:  Revvity Health Sciences Inc L0067RED
Encompass
Tag-lite receptor binding assays use either a green or a red fluorescent ligand to function. This Adenosine A1 receptor red antagonist is a XAC derivative labeled with a red emitting HTRF fluorescent probe. This ligand is best suited for use in the Adenosine A1 receptorbinding assay.
Catalog No. 50-211-9175
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.