Learn More
Adenosine A2aR Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NB3005970.025ML
Description
Adenosine A2aR Polyclonal specifically detects Adenosine A2aR in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
Adenosine A2aR | |
Polyclonal | |
Unconjugated | |
PBS, 1 mg/ml BSA with 0.05% Sodium Azide | |
ADORA2A | |
Synthetic peptide corresponding to residues E(373) S H G D M G L P D V E L L S H E L K(391) of canine A2aAR. | |
Affinity Purified | |
RUO | |
135 | |
Human | |
IgG |
Western Blot | |
0.69 mg/mL | |
Western Blot 1:1000, Simple Western 1:20, Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:20-1:200 | |
adenosine A2 receptor, adenosine A2a receptor, adenosine receptor A2a, adenosine receptor subtype A2a, ADORA 2, ADORA2, hA2aR, RDC8 | |
Rabbit | |
45 kDa | |
0.025 mL | |
Primary | |
Detects adenosine receptor A2a. This does not detect other AR subtypes. | |
Store at -20°C. Avoid freeze/thaw cycles. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only