Learn More
Revvity Health Sciences Inc Adenosine A2BR fluorescent probe, 1 Unit
Supplier: Revvity Health Sciences Inc L0068RED
Tag-lite receptor binding assays use either a green or a red fluorescent ligand to function. This Adenosine A2B receptor red antagonist is a XAC derivative labeled with a red emitting HTRF fluorescent probe. This ligand is best suited for use in the Adenosine A2B receptorbinding assay.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.