Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenosine Deaminase/ADA Antibody (SR1685), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322209100UL
Description
Adenosine Deaminase/ADA Monoclonal antibody specifically detects Adenosine Deaminase/ADA in Human, Rat samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
Adenosine Deaminase/ADA | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Western Blot, Immunofluorescence, Immunoprecipitation | |
SR1685 | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
ADA1, adenine deaminase, Adenosine aminohydrolase, adenosine deaminase, EC 3.5.4.4 | |
A synthesized peptide derived from human Adenosine Deaminase/ADA (Uniprot #: P00813) | |
100 μg | |
Cancer, Endocrinology | |
100 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction