Learn More
Crystal Chem Inc Adenosine Deaminase (ADA) Kit, 96-tests
Supplier: Crystal Chem Inc 80249
The Adenosine Deaminase Assay is used to quantify levels of human adenosine deaminase in human serum and plasma using only a 5 µL sample volume. Highlights: Extended linearity from 0 - 200 U/L. Specific for ADA and has no detectable reaction with other nucleosides. Requires as little as 5 µL, ideal for research applications. Stable liquid stable format requires no reagent preparation. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.