Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Adenosine Deaminase (Calf)

Catalog No. 89932664 Shop All Abnova Corporation Products
Click to view available options
:
250 Units

Used for Enzyme

#REF!

Specifications

For Use With (Application) Enzyme Activity
Format Lyophilized
Molecular Weight (g/mol) ∽33kDa
Name Adenosine Deaminase (Calf)
Preparation Method Native protein chromatographically purified from Calf Spleen by a modification of the method of Pfrogner, Arch. Biochim. Biophys., 119, 141 (1967)
Quantity 250 Units
Storage Requirements Store at 4°C.
Regulatory Status RUO
Common Name ADENOSINE
Assays Adenosine Deaminase Assay
Biological Activity ≥15 units per mg dry weight. One Unit converts one micromole of adenosine to inosine per minute at 25°C, pH 7.4.
Host Species Bovine
Protein Tag None
Expression System Native protein chromatographically purified from Calf Spleen by a modification of the method of Pfrogner, Arch. Biochim. Biophys., 119, 141 (1967)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.