Learn More
Adenosine Deaminase Rabbit anti-Zebrafish, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572657
Description
Catalyzes the hydrolytic deamination of adenosine and 2-deoxyadenosine. Plays an important role in purine metabolism and in adenosine homeostasis. Modulates signaling by extracellular adenosine, and so contributes indirectly to cellular signaling events. May act as a positive regulator of T-cell coactivation (By similarity).Specifications
Adenosine Deaminase | |
Polyclonal | |
Unconjugated | |
ADA | |
ada; ADA1; adenosine aminohydrolase; Adenosine deaminase; RP11-61L14.5; zgc:92028 | |
Rabbit | |
Protein A | |
RUO | |
436919 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.52 mg/mL | |
PBS with 0.09% sodium azide | |
Q6DG22 | |
ADA | |
KLH conjugated synthetic peptide between 114-148 amino acids of DANRA ADA | |
400 μL | |
Primary | |
Zebrafish | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.