Learn More
Adenosine Deaminase Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA554912
Description
Immunogen sequence: QVELHVHLDG SIKPETILYY GRRRGIALPA NTAEGLLNVI GMDKPLTLPD FLAKFDYYMP AIAGCREAIK RIAYEFVEMK AKEGVVYVEV RYSPHLLANS KVEPIPWNQA EGDLTPDEVV ALVGQGLQEG ERDFGVKARS ILCC Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 88%.
Catalyzes the hydrolytic deamination of adenosine and 2-deoxyadenosine. Plays an important role in purine metabolism and in adenosine homeostasis. Modulates signaling by extracellular adenosine, and so contributes indirectly to cellular signaling events. Acts as a positive regulator of T-cell coactivation, by binding DPP4. Its interaction with DPP4 regulates lymphocyte-epithelial cell adhesion.Specifications
Adenosine Deaminase | |
Polyclonal | |
Unconjugated | |
ADA | |
Liquid | |
ADA | |
Recombit protein corresponding to Human Adenosine Deaminase | |
Antigen affinity chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P00813 | |
RP11-61L14.5, ADAR1, DRADA, DSH, DSRAD, G1P1, IFI-4, IFI4, K88DSRBP, P136, 136 kDa double-stranded RNA-binding protein, adenosine deaminase acting on RNA 1-A, double-stranded RNA-specific adenosine deaminase, dsRNA adenosine deaminase, interferon-induced protein 4, interferon-inducible protein 4 | |
Rabbit | |
IgG | |
100 μL | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Primary | |
100 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research Use Only