Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibson Bioscience Reagents: KOH 40%

Catalog No. 21200316
Click to view available options
:

For demonstration of yeast and fungal elements in wet clinical preparations

Specifications

For Use With (Application)

Demonstration of yeast and fungal elements in wet clinical preparations

Product Type

KOH 40%

Formulation

Potassium hydroxide 400g, deionized water 1L

Storage Requirements

2° to 25°C

Test Time

10 min.

Volume (Metric)

50mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.