Learn More
ADFP Mouse anti-Human, Clone: ADFP/1493, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB14486
Description
Mouse monoclonal antibody raised against partial recombinant human ADFP.
- High specificity
- High reactivity
Specifications
ADFP | |
Monoclonal | |
Unconjugated | |
Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
Q99541 | |
ADFP | |
Recombinant protein corresponding to amino acids 249-376 of human ADFP. | |
100 μg | |
Primary | |
Human | |
Liquid |
Immunofluorescence, Immunohistochemistry (PFA fixed) | |
ADFP/1493 | |
Mouse monoclonal antibody raised against partial recombinant human ADFP. | |
In 10 mM PBS (0.05% BSA, 0.05% sodium azide). | |
ADRP/MGC10598 | |
Mouse | |
Protein A/G purification | |
RUO | |
123 | |
Store at 4°C. | |
IgG2b λ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
For Research Use Only (RUO)