Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO Advanced MR Legal for Trade Analytical Balance

METTLER TOLEDO Advanced MR Legal for Trade Analytical Balances offer a highly precise weigh cell with FACT automatic internal adjustment.

Supplier:  METTLER TOLEDO 30666210

 View more versions of this product

Catalog No. 01-804-279


missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart

Description

Description

User management within the 4.5-inch touchscreen increases efficiency of daily tasks with 8 built-in applications and statistical data analysis. Designed with full metal housing and overload protection, these balances will provide high quality performance for years to come. When your balance is not in use, enable the power-saving mode to reduce energy consumption and save costs.

MR Analytical Balances include:

  • FACT automatic internal adjustment
  • OIML/NTEP Approved
  • 4.5 inch color touchscreen
  • 8 built-in applications
  • Statistical data analysis
  • User management
  • Guided leveling
  • 4 interfaces: USB-A, USB-C, RS232, Ethernet
  • Bluetooth option
  • Full metal housing
  • Overload protection

The MR Balance is the successor to the ML or ME-T Balance. See how it ranks among our balance lines:

  • XPR Excellence Analytical Balances
  • MX Advanced Analytical Balances
  • MR Advanced Analytical Balances
  • MA Standard Analytical Balances
  • ME Standard Analytical Balances
  • LA Standard Analytical Balances
Specifications

Specifications

USB-A, USB-C, RS232, Ethernet
Color Touchscreen
Metal
2 sec.
100/240 V, 50/60 Hz
320 g
0.1 mg
MR304/A
Analytical Balance
Class I
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.