Learn More
Gold Biotechnology Inc AEBSF, 30827-99-7, MFCD00132962, 5 g
Supplier: Gold Biotechnology Inc A5405
Formula: C8H10FNO2S · HCl, MW: 239.69 g/mol, Store at -20°C, Protect from light, AEBSF is an irreversible serine protease inhibitor.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.