Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aegeline 98.0+%, TCI America™
SDP

Catalog No. p-7118242 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 456-12-2
Molecular Formula C18H19NO3
Molecular Weight (g/mol) 297.35
MDL Number MFCD00048045
InChI Key QRFDENJATPJOKG-UHFFFAOYNA-N
Synonym Aegelin, (E)-N-[2-Hydroxy-2-(4-methoxyphenyl)ethyl]cinnamamide
PubChem CID 15558419
IUPAC Name N-[2-hydroxy-2-(4-methoxyphenyl)ethyl]-3-phenylprop-2-enamide
SMILES COC1=CC=C(C=C1)C(O)CNC(=O)C=CC1=CC=CC=C1

Specifications

Melting Point 180°C
Color White-Yellow
Quantity 5 g
Formula Weight 297.35
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Aegeline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.