Learn More
Revvity Health Sciences Inc AequoZen FroZen cells, Dopamine D2L, Human Recombinant, CHO-K1
Supplier:  Revvity Health Sciences Inc ES171AF
Encompass
Frozen Cells validated for calcium testing: Dopamine D2L receptor, human recombinant, in CHO-K1 host cell. We provide one vial of frozen cells (10 million cells/vial). Some of our Frozen cells may be restricted for sale in specified countries. Please inquire at your local sales office for more information. You are authorized to utilize these frozen cell preparations one time only. Any attempt to transfer, re-use, or propagate these cells is expressly unauthorized and a violation of the product terms and conditions of sale. Features: Pre-validated batches of ready-to-use cells for calcium testing Optimized to alleviate biological variability Step-by-Step protocols for culture-free functional testing on our validated cells Affordable access to small aliquots of cells (minimum size: 10 million cells/vial) Ready-to-use cells for Profiling, Pharmacology or Screening applications Up-scaling and customized developments The AequoScreen technology is a generic GPCR technology which can be use
Catalog No. 50-209-8873
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.