Learn More
Sigma Aldrich Fine Chemicals Biosciences Agar, 9002-18-0, MFCD00081288, 100 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 05040100G
Encompass
Linear Formula: (C12H18O9)n, powder, for microbiology, Synonym: Agar-agar, Gum agar. Agar has been used as reagent for the following studies: Plant cell culture studies. As medium supplement for the growth of wild-type Arabidopsis thaliana (ecotype Columbia) and transgenic seeds. For the solidification of culture medium of cork oak somatic embryos. It may be used for incubation of single antibody-forming cell to investigate the plaque formation. Agar suspension culture may be used for the analysis of cell transformation of palyoma virus.
Catalog No. NC0684390
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.