Learn More
Across International Ai -40°C 4.6L Capacity 316L SST Cold Trap w/ KF25 Ports ETL
SDP
Supplier:  Across International T40XUL
Encompass_Preferred
The new Ai 3rd Gen -40C/F T40 cold traps all come ETL certified, standard. They have a small footprint, but at the same time, offering you the same cooling power. New features include a large 316L SST 4.6 liter trap, 316L corrosion-resistant trapping layers and vacuum connections, PTFE encapsulated silicone sealing gasket, upward KF25 vacuum connections provide you safer and easier vacuum tubing connection and removal. No chilling fluid required, just plug and play.
Catalog No. 50-225-1300
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.