Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIM 100, Tocris Bioscience™

Catalog No. p-4835928 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective Ack1 (TNK2) inhibitor

Potent Ack1 (TNK2) inhibitor (IC50 = 22 nM). Exhibits selectivity for Ack1 over ABL1, BTK, Lck and LYN; exhibits no inhibition of 25 other kinases. Suppresses phosphorylation of Tyr267 of the androgen receptor in prostate cancer cells. Also suppresses growth of radioresistant castration-resistant prostate cancer in xenograft tumors in mice.

Chemical Identifiers

CAS 873305-35-2
Molecular Formula C23H21N3O2
Molecular Weight (g/mol) 371.44
InChI Key XNFHHOXCDUAYSR-SFHVURJKSA-N
Synonym s-5,6-diphenyl-n-tetrahydrofuran-2-yl methyl furo 2,3-d pyrimidin-4-amine, 5,6-diphenyl-4-s-tetrahydrofuran-2-yl-methyl amino furo 2,3-d pyrimidine, 5,6-diphenyl-n-2s-tetrahydro-2-furanyl methyl furo 2,3-d pyrimidin-4-amine, 5,6-diphenyl-n-2s-tetrahydrofuran-2-yl methyl furo 2,3-d pyrimidin-4-amine, n-2s-oxolan-2-ylmethyl-5,6-diphenylfuro 2,3-d pyrimidin-4-amine
PubChem CID 11501591
IUPAC Name N-[[(2S)-oxolan-2-yl]methyl]-5,6-diphenylfuro[2,3-d]pyrimidin-4-amine
SMILES C1CC(OC1)CNC2=C3C(=C(OC3=NC=N2)C4=CC=CC=C4)C5=CC=CC=C5

Specifications

Quantity 10 mg
Formula Weight 371.43
Chemical Name or Material AIM 100
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.