Learn More
AIMS™ CO2 Chamber
For mice and rats
$39.30 - $1217.00
Catalog Number | Mfr. No. | Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Includes | Price | Quantity & Availability | ||||
14-370-181
|
AIMS™
COBL |
Liner Bag | Disposable Chamber-Liner Bags |
Each for $39.30
|
|
||||
14-370-180
|
AIMS™
COIC |
CO2 Chamber | Acrylic lid with door, 7 ft. tubing, 50 disposable insert bags |
Each for $1,217.00
|
|
||||
Description
Simple, quick and easy to use
- Easy to see animals in chamber with see-through acrylic lid with door and quick connect with valve, 2.13m (7 ft.) tubing
- 50 disposable chamber-liner bags collect animal feces and urine, keeping the chamber clean
- 22.86cm (9 in.) diameter, easy to keep on bench
- Allows user to place animals in chamber without losing CO2
- Delivery tube within chamber fills chamber from bottom, decrease induction time – more humane; provides for ultra-quick animal induction
- Solid sides prevent viewing of animals during processes
Order Info
Replacement disposable chamber-line bags available separately
Specifications
Single-use |