Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ AIN-76A Semipurified Diet

Catalog No. p-7236589 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:

AIN-76A Semipurified Diet is intended for growth and maintenance of animals during the first year of life.

  • Contains casein purified high nitrogen, DL-methionine, sucrose, corn starch, Alphacel (non-nutritive bulk), corn oil, choline bitartrate, AIN mineral mix, and a 10-fold increase in the amount of vitamin K source to 500 μg/kg.

Specifications

Format

Powder

Quantity

10 kg

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.