Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ AIN-93M Diet, Powder

AIN-93M Diet, Powder can be used for maintenance of adult rodents.

$976.00 - $3574.00

Specifications

Product Type Animal Diet
Components Casein 14%, Dextrinized Cornstarch 15.5%, Sucrose 10%, Corn Starch 46.5692%, Alphacel, Non-Nutritive Bulk 5%, Soybean Oil 4%, AIN-93M Mineral Mix 3.5%, L-Cystine 0.18%, AIN-93-VX Vitamin Mix 1%, Choline bitartrate 0.25%, tert-Butylhydroquinone 0.0008%
Format Powder
Species Rodent
Storage Requirements 4°C
View More Specs

Products 4
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
ICN96039610
View Documents
MP Biomedicals™
0296039610
10 kg
Each for $976.00
Add to Cart
 
ICN96039620
View Documents
MP Biomedicals™
0296039620
20 kg
Each for $1,633.00
Add to Cart
 
02-960-39630
View Documents
MP Biomedicals™
0296039630
30 kg
Each for $2,061.00
Add to Cart
 
ICN96039650
View Documents
MP Biomedicals™
0296039650
50 kg
Each for $3,574.00
Add to Cart
 
Description

Description

  • Revised American Institute of Nutrition Rodent Diets AIN-76 and AIN-76A formulations to support maintenance of adult animals.
Specifications

Specifications

Animal Diet
Powder
4°C
Casein 14%, Dextrinized Cornstarch 15.5%, Sucrose 10%, Corn Starch 46.5692%, Alphacel, Non-Nutritive Bulk 5%, Soybean Oil 4%, AIN-93M Mineral Mix 3.5%, L-Cystine 0.18%, AIN-93-VX Vitamin Mix 1%, Choline bitartrate 0.25%, tert-Butylhydroquinone 0.0008%
Rodent
Videos
SDS
Documents

Documents

Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.