Learn More
TARGETMOL CHEMICALS INC AKOS037652256 5MG
Supplier:  TARGETMOL CHEMICALS INC T72073
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. AKOS037652256 can be used as a TRPML modulator for the treatment of diseases associated with TRPML activity such as lysosomal accumulation disorders muscular dystrophy common age-related neurodegenerative diseases oxidative stress or reactive oxygen species (ROS)-related diseases and ageing. purity: 98%
Catalog No. 50-257-9247
missing translation for 'QuantityLeft'