Learn More
TARGETMOL CHEMICALS INC ALBIFLORIN 20MG
Supplier:  TARGETMOL CHEMICALS INC T6S1653
Encompass_Preferred
Also available in 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Albiflorin (Alibiflorin) has anticonvulsant effect for pain relief. 2. Albiflorin and paeoniflorin has neuroprotective action against Glu toxicity. 3. Albiflorin and paeoniflorin significantly ameliorated Glu-induced reduction of cell viability nuclear and mitochondrial apoptotic alteration reactive oxygen species accumulation and B-cell lymphoma 2 (Bcl-2)/Bax ratio. 4. Albiflorin can serve as a new chemical marker for the quality control of Paeoniae Radix and the Chinese Pharmacopoeia can be updated via has similar anti-inflammatory effects to paeoniflorin. purity: 99%
Catalog No. 50-257-6377
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.