Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Albumin, from Eggs
SCI_ED

Catalog No. p-4416313 Shop All Fisherbrand Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Reagent Grade

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 9006-59-1
Molecular Formula C16H26O5
Molecular Weight (g/mol) 298.38
MDL Number MFCD00130424
InChI Key UOXVFQCRPDLSFN-UHFFFAOYNA-N
Synonym 3,4-dihydroxy-2-methoxy-4-methyl-3-2-methyl-3-3-methyl-but-2-enyl-oxiranyl-cyclohexanone, ova, 2s,3r,4r-3,4-dihydroxy-2-methoxy-4-methyl-3-2s,3r-2-methyl-3-3-methylbut-2-en-1-yl oxiran-2-yl cyclohexan-1-one, 2s,3r,4r-3,4-dihydroxy-2-methoxy-4-methyl-3-2s,3r-2-methyl-3-3-methylbut-2-enyl oxiran-2-yl cyclohexan-1-one
PubChem CID 5289086
IUPAC Name 3,4-dihydroxy-2-methoxy-4-methyl-3-[2-methyl-3-(3-methylbut-2-en-1-yl)oxiran-2-yl]cyclohexan-1-one
SMILES COC1C(=O)CCC(C)(O)C1(O)C1(C)OC1CC=C(C)C

Specifications

Color Yellow
Density 1g/mL
Packaging Reagent Grade
Quantity 25 g
Formula Weight 33.8 to 46.1
Grade Reagent
Chemical Name or Material Albumin, from Eggs
ShelfLife 1 year
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.