Learn More
MP Biomedicals™ Albumin, Human, Fraction V Powder, 10g Lyoph.
Shop All MP Biomedicals, Inc ProductsSpecifications
For Use With (Application)
Immunoassay, Mammalian Cell Culture
Name
Human Albumin Fraction V
pH Range
6.8
Preparation Method
Human Protein is prepared by a modification of the Cohn procedure
Quantity
10 g
Source
Plasma
Storage Requirements
2° to 8°C
Form
Lyophilized
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.