Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alconox™ Tergazyme™ Enzyme-Active Powered Detergent

Concentrated, anionic detergent with protease enzyme for manual and ultrasonic cleaning

Supplier:  Alconox™ 1350

 View more versions of this product

Catalog No. 16-000-117


missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart

Description

Description

Tergazyme consists primarily of a homogeneous blend of sodium linear alkylaryl sulfonate, phosphates, carbonates, and protease enzyme.

  • Concentrated, biodegradable and disposable
  • Ideal for removal of proteinaceous soils, tissue, blood and body fluids from glassware, metals, plastic, ceramic, porcelain, rubber and fiberglass
  • Replaces corrosive acids and hazardous solvents
  • Free rinsing gives reliable results without interfering residues
  • Dilute 1:100 for typical use (10g in 1L of water)
  • Phosphate Content: 7.5% (as Phosphorus)
  • Use as cleaning agent in Reverse Osmosis and Ultra-Filtration Systems
  • Use to pass cleaning validation tests for lab accreditation and plant inspection approval
  • USDA authorized for use in federally inspected meat and poultry plants

Used to clean:
Medical instruments/devices, dairy equipment, laboratory ware, reverse osmosis and ultrafiltration membranes and units, sampling apparatus, pharmaceutical apparatus, cosmetics manufacturing equipment, tubing, pipes, optical parts, process equipment, industrial parts, desalination plants, tanks and reactors. Passes inhibitory residue test for water analysis.

Used to remove:
Soil, grit, grime, blood, tissue, grease, fats, oils, proteinaceous soils, dairy proteins, particulates, solvents and bioreactor residue.

Surfaces cleaned:
Corrosion inhibited formulation recommended for glass, metal, stainless steel, porcelain, ceramic, plastic, rubber and fiberglass. Can be used on soft metals such as copper, aluminum, zinc and magnesium if rinsed promptly.

Cleaning method:
Soak, brush, sponge, cloth, ultrasonic, flow through clean-inplace. Will foam—not for spray or machine use.

Shelf life: 2 years from date of manufacture.

Order Info

0.5kg (1 lb.) makes 49.2L (13 gal.) cleaning solution.

Specifications

Specifications

Enzyme-Active Powered Detergent
9.5
Carton
Remove proteinaceous soils, tissue, blood and body fluids
22.6 kg
Powder
White, Cream
Store closed and upright in a cool dry place at 15° to 30°C.
50 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.