Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALK Mouse anti-Human, Clone: ALK/1503, Abnova™

Mouse Monoclonal Antibody

Supplier:  Abnova Corporation MAB14536

Catalog No. 89-034-619


missing translation for 'QuantityLeft'
Add to Cart

Description

Description

Mouse monoclonal antibody raised against partial recombinant human ALK.

  • High specificity
  • High reactivity
Specifications

Specifications

ALK
ALK/1503
Mouse monoclonal antibody raised against partial recombinant human ALK.
In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Liquid
ALK
Recombinant protein corresponding to amino acids 1360-1460 of human ALK.
Protein A/G purification
RUO
Primary
238
Flow Cytometry, Immunofluorescence, Western Blot
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Q9UM73
CD246/Ki-1/TFG/ALK
Mouse
IgG1, kappa
100 ug
Store at 4°C.
Monoclonal
Human
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

For Research Use Only (RUO)