Learn More
Alkaline-Iodide-Azide, Pomeroy for Dissolved Oxygen Analysis, Reagents
Alkaline-Iodide-Azide, Pomeroy for Dissolved Oxygen Analysis, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$378.50 - $1531.00
Specifications
Color | Colorless |
---|---|
Volume (Metric) | 1 L |
Packaging | Poly Bottle |
Sterility | Non-sterile |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
01-336-041
|
Reagents Holdings Llc
CA0146001A |
1 L |
Each for $703.50
|
|
|||||
01-336-040
|
Reagents Holdings Llc
CA0146004A |
4 L |
Each for $1,531.00
|
|
|||||
01-336-039
|
Reagents Holdings Llc
CA014600500A |
500 mL |
Each for $378.50
|
|
|||||
Specifications
Colorless | |
1 L | |
Poly Bottle | |
Non-sterile |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.