Learn More
Alkalinity Titrant, Low, AquaPhoenix Scientific™
SCI_ED
Supplier:  Aqua Phoenix Scientific SA1555P
Catalog No. S88720
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Description
Description
Specifications
Specifications
1 drop = 10 ppm as CaCO3/25 mL
Sulfuric Acid 0.12 N
500 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.