Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

all cis-5,8,11,14,17-Eicosapentaenoic Acid 95.0+%, TCI America™
SDP

Catalog No. p-7128997 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mg
500 mg

Chemical Identifiers

CAS 10417-94-4
Molecular Formula C20H30O2
Molecular Weight (g/mol) 302.46
MDL Number MFCD00065716
InChI Key JAZBEHYOTPTENJ-JLNKQSITSA-N
Synonym EPA, all cis-5,8,11,14,17-Icosapentaenoic Acid
PubChem CID 446284
ChEBI CHEBI:28364
IUPAC Name (5Z,8Z,11Z,14Z,17Z)-icosa-5,8,11,14,17-pentaenoic acid
SMILES CC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CCCC(O)=O

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 302.46
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material all cis-5,8,11,14,17-Eicosapentaenoic Acid
EINECSNumber (2)-0609
TSCA No
Recommended Storage Freezer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.