Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

all cis-8,11,14-Eicosatrienoic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7129083 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
10 mg
50 mg

Chemical Identifiers

CAS 1783-84-2
Molecular Formula C20H34O2
Molecular Weight (g/mol) 306.49
MDL Number MFCD00065721
InChI Key HOBAELRKJCKHQD-QNEBEIHSSA-N
Synonym dihomo-gamma-linolenic acid, dgla, gamma-homolinolenic acid, 8,11,14-eicosatrienoic acid, homo-gamma-linolenic acid, cis-8,11,14-eicosatrienoic acid, bishomo-gamma-linolenic acid, z,z,z-8,11,14-eicosatrienoic acid, all-cis-8,11,14-eicosatrienoic acid, 8z,11z,14z-icosa-8,11,14-trienoic acid
PubChem CID 5280581
ChEBI CHEBI:53486
IUPAC Name (8Z,11Z,14Z)-icosa-8,11,14-trienoic acid
SMILES CCCCCC=CCC=CCC=CCCCCCCC(=O)O

Specifications

Color Yellow
Boiling Point 200°C
Quantity 50 mg
Formula Weight 306.49
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material all cis-8,11,14-Eicosatrienoic Acid
EINECSNumber (2)-0609
RTECSNumber JX3880000
TSCA No
Recommended Storage Freezer (-20 °C)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.