Learn More
Beckman Coulter Beckman Coulter Allegra X-30 Cell Culture Pkg
Supplier:  Beckman Coulter B08536
Encompass_Preferred
Cell Culture Application Package: Allegra X-30, IVD, 120 V, 60 Hz, Ambient Benchtop Centrifuge , 4x 400mL SX4400 Swinging-bucket Rotor,15 mL Conical Adapters (2 sets of 2), 50 mL Conical Adapters (2 sets of 2) Max. tube capacity: 15 mL = 20 tubes / 50 mL = 12 tubes
Catalog No. 50-163-1958
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.