Learn More
3M™ Almond Protein Rapid Kit L25ALM, Sold by AquaPhoenix Scientific
Shop All AquaPhoenix Scientific Products
- Results visible at 11 ± 1 min. after application of sample
- Rapid, qualitative immunochromatographic lateral flow test cartridges designed for detection of allergenic food proteins
- Designed to detect both processed and unprocessed allergen proteins
- Test can accommodate environmental swab, CIP rinse water and food samples
Specifications
Format
Kit
Product Type
Microbiology Media
Test Type
Allergen
Quantity
25/Cs.