Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell AlphaTec™ 1500 PLUS - Lab Coat Model 208

Catalog No. 17000263 Shop All Ansell Products
Click to view available options
Size:

Breathable lab coats are made of anti-static SMS material suitable for electro-static sensitive and manufacturing work environments.

  • Short style, white lab coats
  • SMS fabric construction
  • Anti-static treatment
  • Air and water vapor permeable
  • Silicone-free, bound seams and low-linting
  • Elastic cuffs, studded front-fastening and left-breast & right-thigh integral pockets

Specifications

Product Type

Controlled Environments Lab Coat

Size

5X-Large

Gender

Unisex

Material

SMS

Color

White

Closure Type

Press Studs

Cuff Style

Elastic

Resistance

Static

No. of Pockets

2

Collar Type

Serged

Seam Type

Bound

Certifications/Compliance (Europe)

CE CATEGORY III (Type PB [6])

For Use With (Application)

Unpacking Bulk Goods, Loading and Unloading Processing Equipment, Production Line Support and Maintenance, Assembly and Inspection of Components

No. per Pack

70

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.