Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ansell AlphaTec™ 1500 PLUS - Lab Coat Model 208
Shop All Ansell Products
Click to view available options
Size:
Small
Medium
Large
X-Large
3X-Large
4X-Large
5X-Large
Description
- Short style, white lab coats
- SMS fabric construction
- Anti-static treatment
- Air and water vapor permeable
- Silicone-free, bound seams and low-linting
- Elastic cuffs, studded front-fastening and left-breast & right-thigh integral pockets
Specifications
Specifications
Product Type | Controlled Environments Lab Coat |
Size | Small |
Gender | Unisex |
Material | SMS |
Color | White |
Resistance | Static |
Collar Type | Serged |
Certifications/Compliance (Europe) | CE CATEGORY III (Type PB [6]) |
For Use With (Application) | Unpacking Bulk Goods, Loading and Unloading Processing Equipment, Production Line Support and Maintenance, Assembly and Inspection of Components |
No. per Pack | 70 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction