Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ Alphatec™ MICROCHEM™ 68-3000 Yellow Coveralls with Hood
SCI_ED

Catalog No. p-6904065 Shop All Ansell Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
This item is not returnable. View return policy
This product contains natural rubber latex which may cause allergic reactions.
This item is not returnable. View return policy
This product contains natural rubber latex which may cause allergic reactions.

Provide an extremely effective barrier against both chemical and biological hazards. Ansell™ Alphatec™ MICROCHEM™ 68-3000 Yellow Coveralls with Hood are durable and multi-layered, and are some of the most comfortable chemical protective materials available.

  • Protection:  Multi-layer barrier fabric effective against numerous chemicals
  • Highly visible:  Bright yellow for improved worker safety
  • Anti-static:  Tested according to EN1149-5 & AATCC76 (8EMSL test method)
  • Design:  Elastic waist, wrists and ankles, double zip closure, double cuffs, ultrasonically welded seams, finger loops, 2 piece respirator fit hood
  • Applications:  Chemicals, oil and petrochemicals, pharmaceutical, food indsutry, sewage purification installations, industrial and tank cleaning, mining
  • Sizes:  Available in Small through 5X-Large
  • Certifications:  EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B
  • Storage:  5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B

Specifications

Size Small
Material Barrier laminate fabric
Color Yellow
Waist Style Elastic
Ankle Style Elastic
Cuff Style Double, Elastic
Neck Style Hooded
Hood Type 3-panel, Respirator Fit
Closure Type Double zipper
Seam Type Welded
Protection Class Liquid and spray tight chemical and biological protection
Latex Free No
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.