Learn More
JDM SCIENTIFIC LLC AlteCap™ Septum capper
Semi-automated septum capper for all manufacturers septum cap mats with tubes in SBS racks, 24, 48 and 96 well plates. Includes: manual, power cable and support adapters for racks and plates to ensure optimal sealing.