Learn More
JDM SCIENTIFIC LLC AlteCap™ Solo - Internal thread Micronic screw caps (96 SBS format)
Semi-automated single decapper. Electric capping & decapping. Compatible with 96 SBS rack format Micronic internal thread screw caps Includes: Manual, stand, USB charging cable & cap driver.