Learn More
JDM SCIENTIFIC LLC AlteCap™ Switch Cassette 12/96 - Internal thread LVL screw caps
Supplier:  JDM SCIENTIFIC LLC 404031
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
AlteCap™ Switch cap driver cassette - 12 channel. Compatible with 96 SBS rack format LVL internal thread screw caps - part number IT96-xxx-BU-P-L
Catalog No. 50-271-9983